Merck
  • Quiescence enables unrestricted cell fate in naive embryonic stem cells.
Quiescence enables unrestricted cell fate in naive embryonic stem cells.
Nature communications (2024-02-27)
Le Tran Phuc Khoa, Wentao Yang, Mengrou Shan, Li Zhang, Fengbiao Mao, Bo Zhou, Qiang Li, Rebecca Malcore, Clair Harris, Lili Zhao, Rajesh C Rao, Shigeki Iwase, Sundeep Kalantry, Stephanie L Bielas, Costas A Lyssiotis, Yali Dou
ABSTRACT
Quiescence in stem cells is traditionally considered as a state of inactive dormancy or with poised potential. Naive mouse embryonic stem cells (ESCs) can enter quiescence spontaneously or upon inhibition of MYC or fatty acid oxidation, mimicking embryonic diapause in vivo. The molecular underpinning and developmental potential of quiescent ESCs (qESCs) are relatively unexplored. Here we show that qESCs possess an expanded or unrestricted cell fate, capable of generating both embryonic and extraembryonic cell types (e.g., trophoblast stem cells). These cells have a divergent metabolic landscape comparing to the cycling ESCs, with a notable decrease of the one-carbon metabolite S-adenosylmethionine. The metabolic changes are accompanied by a global reduction of H3K27me3, an increase of chromatin accessibility, as well as the de-repression of endogenous retrovirus MERVL and trophoblast master regulators. Depletion of methionine adenosyltransferase Mat2a or deletion of Eed in the polycomb repressive complex 2 results in removal of the developmental constraints towards the extraembryonic lineages. Our findings suggest that quiescent ESCs are not dormant but rather undergo an active transition towards an unrestricted cell fate.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-5-methylcytosine Antibody, clone 33D3, clone 33D3, from mouse
Sigma-Aldrich
c-Myc Inhibitor, The c-Myc Inhibitor, also referenced under CAS 403811-55-2, controls the biological activity of c-Myc.
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
PD 0325901, ≥98% (HPLC)
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit