Merck
  • Nonradioactive, ultrasensitive site-specific protein-protein photocrosslinking: interactions of alpha-helix 2 of TATA-binding protein with general transcription factor TFIIA and transcriptional repressor NC2.
Nonradioactive, ultrasensitive site-specific protein-protein photocrosslinking: interactions of alpha-helix 2 of TATA-binding protein with general transcription factor TFIIA and transcriptional repressor NC2.
Nucleic acids research (2008-10-01)
Younggyu Kim, Yon W Ebright, Adam R Goodman, Danny Reinberg, Richard H Ebright
ABSTRACT
We have developed an approach that enables nonradioactive, ultrasensitive (attamole sensitivity) site-specific protein-protein photocrosslinking, and we have applied the approach to the analysis of interactions of alpha-helix 2 (H2) of human TATA-element binding protein (TBP) with general transcription factor TFIIA and transcriptional repressor NC2. We have found that TBP H2 can be crosslinked to TFIIA in the TFIIA-TBP-DNA complex and in higher order transcription-initiation complexes, and we have mapped the crosslink to the 'connector' region of the TFIIA alpha/beta subunit (TFIIAalpha/beta). We further have found that TBP H2 can be crosslinked to NC2 in the NC2-TBP-DNA complex, and we have mapped the crosslink to the C-terminal 'tail' of the NC2 alpha-subunit (NC2alpha). Interactions of TBP H2 with the TFIIAalpha/beta connector and the NC2alpha C-terminal tail were not observed in crystal structures of TFIIA-TBP-DNA and NC2-TBP-DNA complexes, since relevant segments of TFIIA and NC2 were not present in truncated TFIIA and NC2 derivatives used for crystallization. We propose that interactions of TBP H2 with the TFIIAalpha/beta connector and the NC2alpha C-terminal tail provide an explanation for genetic results suggesting importance of TBP H2 in TBP-TFIIA interactions and TBP-NC2 interactions, and provide an explanation-steric exclusion-for competition between TFIIA and NC2.
Sigma-Aldrich
2-Nitro-5-thiocyanatobenzoic acid, powder
Sigma-Aldrich
Avidin−Agarose from egg white, aqueous glycerol suspension
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Triethylsilane, 97%
Sigma-Aldrich
Triethylsilane, 99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
2,2′-Dithiodipyridine, powder
Sigma-Aldrich
2,2′-Dithiodipyridine, ≥99.0% (GC)