Merck
A membrane-associated MHC-I inhibitory axis for cancer immune evasion.
Cell (2023-08-10)
Xufeng Chen, Qiao Lu, Hua Zhou, Jia Liu, Bettina Nadorp, Audrey Lasry, Zhengxi Sun, Baoling Lai, Gergely Rona, Jiangyan Zhang, Michael Cammer, Kun Wang, Wafa Al-Santli, Zoe Ciantra, Qianjin Guo, Jia You, Debrup Sengupta, Ahmad Boukhris, Hongbing Zhang, Cheng Liu, Peter Cresswell, Patricia L M Dahia, Michele Pagano, Iannis Aifantis, Jun Wang
ABSTRACT
Immune-checkpoint blockade has revolutionized cancer treatment, but some cancers, such as acute myeloid leukemia (AML), do not respond or develop resistance. A potential mode of resistance is immune evasion of T cell immunity involving aberrant major histocompatibility complex class I (MHC-I) antigen presentation (AP). To map such mechanisms of resistance, we identified key MHC-I regulators using specific peptide-MHC-I-guided CRISPR-Cas9 screens in AML. The top-ranked negative regulators were surface protein sushi domain containing 6 (SUSD6), transmembrane protein 127 (TMEM127), and the E3 ubiquitin ligase WWP2. SUSD6 is abundantly expressed in AML and multiple solid cancers, and its ablation enhanced MHC-I AP and reduced tumor growth in a CD8+ T cell-dependent manner. Mechanistically, SUSD6 forms a trimolecular complex with TMEM127 and MHC-I, which recruits WWP2 for MHC-I ubiquitination and lysosomal degradation. Together with the SUSD6/TMEM127/WWP2 gene signature, which negatively correlates with cancer survival, our findings define a membrane-associated MHC-I inhibitory axis as a potential therapeutic target for both leukemia and solid cancers.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Okadaic acid ammonium salt from Prorocentrum concavum, ≥90% (HPLC), solid
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial