Merck
  • Themis controls T cell activation, effector functions, and metabolism of peripheral CD8+ T cells.
Themis controls T cell activation, effector functions, and metabolism of peripheral CD8+ T cells.
Life science alliance (2023-09-23)
Namrata Gautam, Lukasz Wojciech, Jiawei Yap, Yen Leong Chua, Eyan Mw Ding, Don Cn Sim, Alrina Sm Tan, Patricia J Ahl, Mukul Prasad, Desmond Wh Tung, John E Connolly, Giulia Adriani, Joanna Brzostek, Nicholas Rj Gascoigne
ABSTRACT
Themis is important in regulating positive selection of thymocytes during T cell development, but its role in peripheral T cells is less understood. Here, we investigated T cell activation and its sequelae using a tamoxifen-mediated, acute Themis deletion mouse model. We find that proliferation, effector functions including anti-tumor killing, and up-regulation of energy metabolism are severely compromised. This study reveals the phenomenon of peripheral adaptation to loss of Themis, by demonstrating direct TCR-induced defects after acute deletion of Themis that were not evident in peripheral T cells chronically deprived of Themis in dLck-Cre deletion model. Peripheral adaptation to long-term loss was compared using chronic versus acute tamoxifen-mediated deletion and with the (chronic) dLck-Cre deletion model. We found that upon chronic tamoxifen-mediated Themis deletion, there was modulation in the gene expression profile for both TCR and cytokine signaling pathways. This profile overlapped with (chronic) dLck-Cre deletion model. Hence, we found that peripheral adaptation induced changes to both TCR and cytokine signaling modules. Our data highlight the importance of Themis in the activation of CD8+ T cells.
Sigma-Aldrich
Anti-Themis Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer