Merck
  • Adaptable toolbox to characterize Alzheimer's disease pathology in mouse models.
Adaptable toolbox to characterize Alzheimer's disease pathology in mouse models.
STAR protocols (2022-12-07)
Youtong Huang, Greg Lemke
ABSTRACT
Here, we describe a highly adaptable toolbox for characterizing and analyzing molecular and histopathological changes in Alzheimer's disease (AD) mouse models. We detail optimized and streamlined approaches from sample preparation to image analysis to facilitate reproducible analyses. We also describe the extraction and measurement of the soluble Aβ level by sandwich ELISA in the cortex and hippocampus of AD mouse models before and after plaque deposition. Finally, we outline the steps for image quantification and analysis using Imaris and ImageJ. For complete details on the use and execution of this protocol, please refer to Huang et al. (2021).1.
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-RTN3 (R458), from rabbit