Merck
  • ANKRD13a controls early cell-death checkpoint by interacting with RIP1 independent of NF-κB.
ANKRD13a controls early cell-death checkpoint by interacting with RIP1 independent of NF-κB.
Cell death and differentiation (2021-11-29)
Minho Won, Kyeong Ah Park, Sup Kim, Eunjin Ju, Youngbok Ko, Heonjong Yoo, Hyunju Ro, Jaeseob Lee, Junseo Oh, Eun Gyo Lee, Sang Yean Kim, Suk Woo Nam, Han-Ming Shen, Min-Kyung Yeo, Jin Man Kim, Gang Min Hur
ABSTRACT
In TNF signaling, ubiquitination of RIP1 functions as an early cell-death checkpoint, which prevents the spatial transition of the signaling complex from complex-I to death-inducing complex-II. Here, we report that ankyrin repeat domain 13a (ANKRD13a) acts as a novel component of complex-II to set a higher signal threshold for the cytotoxic potential of TNF. ANKRD13a deficiency is sufficient to turn the response to TNF from survival to death by promoting the formation of complex-II without affecting NF-κB activation. ANKRD13a binds to ubiquitinated-RIP1 via its UIM, and subsequently limits the association of FADD and caspase-8 with RIP1. Moreover, high ANKRD13a expression is inversely correlated with apoptotic phenotypes in ovarian cancer tissues and is associated with poor prognosis. Our work identifies ANKRD13a as a novel gatekeeper of the early cell-death checkpoint, which may function as part of an escape mechanism from cell death in some cancers.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
IKK-2 Inhibitor IV, The IKK-2 Inhibitor IV, also referenced under CAS 507475-17-4, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
Necrostatin-1, Necrostatin-1, CAS 4311-88-0, is a cell-permeable, potent, and selective blocker of necroptosis (EC₅₀ = 494 nM in FADD-deficient Jurkat cells treated with TNF-α).
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-A20 Mouse mAb (59A426), liquid, clone 59A426, Calbiochem®