Merck
  • Dynamic de novo heterochromatin assembly and disassembly at replication forks ensures fork stability.
Dynamic de novo heterochromatin assembly and disassembly at replication forks ensures fork stability.
Nature cell biology (2023-07-07)
Vincent Gaggioli, Calvin S Y Lo, Nazaret Reverón-Gómez, Zuzana Jasencakova, Heura Domenech, Hong Nguyen, Simone Sidoli, Andrey Tvardovskiy, Sidrit Uruci, Johan A Slotman, Yi Chai, João G S C Souto Gonçalves, Eleni Maria Manolika, Ole N Jensen, David Wheeler, Sriram Sridharan, Sanjiban Chakrabarty, Jeroen Demmers, Roland Kanaar, Anja Groth, Nitika Taneja
ABSTRACT
Chromatin is dynamically reorganized when DNA replication forks are challenged. However, the process of epigenetic reorganization and its implication for fork stability is poorly understood. Here we discover a checkpoint-regulated cascade of chromatin signalling that activates the histone methyltransferase EHMT2/G9a to catalyse heterochromatin assembly at stressed replication forks. Using biochemical and single molecule chromatin fibre approaches, we show that G9a together with SUV39h1 induces chromatin compaction by accumulating the repressive modifications, H3K9me1/me2/me3, in the vicinity of stressed replication forks. This closed conformation is also favoured by the G9a-dependent exclusion of the H3K9-demethylase JMJD1A/KDM3A, which facilitates heterochromatin disassembly upon fork restart. Untimely heterochromatin disassembly from stressed forks by KDM3A enables PRIMPOL access, triggering single-stranded DNA gap formation and sensitizing cells towards chemotherapeutic drugs. These findings may help in explaining chemotherapy resistance and poor prognosis observed in patients with cancer displaying elevated levels of G9a/H3K9me3.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-H2AK15ub Antibody, clone EDL H2AK15-4, clone EDL H2AK15-4, from mouse