Merck
  • Echinocystic acid induces the apoptosis, and inhibits the migration and invasion of non-small cell lung cancer cells.
Echinocystic acid induces the apoptosis, and inhibits the migration and invasion of non-small cell lung cancer cells.
Medical oncology (Northwood, London, England) (2023-05-19)
Duojie Zhu, Bin Li, Cheng Wang, Peng Jiang, Futian Tang, Yumin Li
ABSTRACT
An increasing amount of evidence has demonstrated the anticancer activity of triterpenes extracted from traditional medicines. Echinocystic acid (EA), a natural triterpene isolated from Eclipta prostrata (L.) L., has previously been shown to exhibit anticancer activity in HepG2 and HL-60 cells. The aim of the present study was to investigate the anticancer activity of EA in non-small cell lung cancer (NSCLC) cells. For this purpose, the viability and proliferation of A549 cells were determined using a Cell Counting Kit-8 and 5-ethynyl-2'-deoxyuridine staining. The migratory and invasive ability of the A549 cells were measured using wound healing and Transwell assays. Hoechst staining was also performed to detect the apoptosis of A549 cells. The proliferation of A549 cells and the distributions of different growth phases were determined using a flow cytometer. Western blot analysis was used to detect the expression levels of cyclin D, partitioning defective 3 homolog (Par3), PI3K, Akt, mTOR, Bax, Bcl-2 and caspase-3. EA inhibited the proliferation, and the migratory and invasive abilities of cultured lung carcinoma cells (A549 cells), and induced cell cycle arrest in the G1 phase of the cell cycle. Treatment with EA upregulated Par3 expression and inhibited the PI3K/Akt/mTOR pathway in vitro. In addition, EA treatment inhibited tumor growth, suppressed proliferation and induced the apoptosis of tumor cells in NSCLC tumor xenografts in mice. On the whole, these results suggest that EA may represent a potential therapeutic agent for NSCLC.
Sigma-Aldrich
Anti-phospho-BCL-2 (pSer87) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-BCL-2 (pSer87) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-mTOR antibody produced in rabbit, 1.0-1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Caspase 3 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-PARD3 antibody produced in mouse, clone 4G5, purified immunoglobulin, buffered aqueous solution
Echinocystic Acid, ≥97%
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-PCNA antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-mTOR (pSer2448) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-PI3K P85 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP62, affinity isolated antibody
Sigma-Aldrich
Anti-Cyclin D Antibody, from rabbit, purified by affinity chromatography