Merck
Membrane lipid remodeling modulates γ-secretase processivity.
The Journal of biological chemistry (2023-02-23)
Edgar Dawkins, Rico J E Derks, Martina Schifferer, Johannes Trambauer, Edith Winkler, Mikael Simons, Dominik Paquet, Martin Giera, Frits Kamp, Harald Steiner
ABSTRACT
Imbalances in the amounts of amyloid-β peptides (Aβ) generated by the membrane proteases β- and γ-secretase are considered as a trigger of Alzheimer's disease (AD). Cell-free studies of γ-secretase have shown that increasing membrane thickness modulates Aβ generation but it has remained unclear if these effects are translatable to cells. Here we show that the very long-chain fatty acid erucic acid (EA) triggers acyl chain remodeling in AD cell models, resulting in substantial lipidome alterations which included increased esterification of EA in membrane lipids. Membrane remodeling enhanced γ-secretase processivity, resulting in the increased production of the potentially beneficial Aβ37 and/or Aβ38 species in multiple cell lines. Unexpectedly, we found that the membrane remodeling stimulated total Aβ secretion by cells expressing WT γ-secretase but lowered it for cells expressing an aggressive familial AD mutant γ-secretase. We conclude that EA-mediated modulation of membrane composition is accompanied by complex lipid homeostatic changes that can impact amyloidogenic processing in different ways and elicit distinct γ-secretase responses, providing critical implications for lipid-based AD treatment strategies.
Sigma-Aldrich
γ-Secretase Inhibitor X, InSolution, ≥90%, 1 mM
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution