Merck
  • Oscillating field stimulation promotes axon regeneration and locomotor recovery after spinal cord injury.
Oscillating field stimulation promotes axon regeneration and locomotor recovery after spinal cord injury.
Neural regeneration research (2021-11-17)
Yi-Xin Wang, Jin-Zhu Bai, Zhen Lyu, Guang-Hao Zhang, Xiao-Lin Huo
ABSTRACT
Oscillating field stimulation (OFS) is a potential method for treating spinal cord injury. Although it has been used in spinal cord injury (SCI) therapy in basic and clinical studies, its underlying mechanism and the correlation between its duration and nerve injury repair remain poorly understood. In this study, we established rat models of spinal cord contusion at T10 and then administered 12 weeks of OFS. The results revealed that effectively promotes the recovery of motor function required continuous OFS for more than 6 weeks. The underlying mechanism may be related to the effects of OFS on promoting axon regeneration, inhibiting astrocyte proliferation, and improving the linear arrangement of astrocytes. This study was approved by the Animal Experiments and Experimental Animal Welfare Committee of Capital Medical University (supplemental approval No. AEEI-2021-204) on July 26, 2021.
Sigma-Aldrich
Anti-Mouse IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution