Merck
  • AT-hook DNA-binding motif-containing protein one knockdown downregulates EWS-FLI1 transcriptional activity in Ewing's sarcoma cells.
AT-hook DNA-binding motif-containing protein one knockdown downregulates EWS-FLI1 transcriptional activity in Ewing's sarcoma cells.
PloS one (2022-10-05)
Takao Kitagawa, Daiki Kobayashi, Byron Baron, Hajime Okita, Tatsuo Miyamoto, Rie Takai, Durga Paudel, Tohru Ohta, Yoichi Asaoka, Masayuki Tokunaga, Koji Nakagawa, Makoto Furutani-Seiki, Norie Araki, Yasuhiro Kuramitsu, Masanobu Kobayashi
ABSTRACT
Ewing's sarcoma is the second most common bone malignancy in children or young adults and is caused by an oncogenic transcription factor by a chromosomal translocation between the EWSR1 gene and the ETS transcription factor family. However, the transcriptional mechanism of EWS-ETS fusion proteins is still unclear. To identify the transcriptional complexes of EWS-ETS fusion transcription factors, we applied a proximal labeling system called BioID in Ewing's sarcoma cells. We identified AHDC1 as a proximal protein of EWS-ETS fusion proteins. AHDC1 knockdown showed a reduced cell growth and transcriptional activity of EWS-FLI1. AHDC1 knockdown also reduced BRD4 and BRG1 protein levels, both known as interacting proteins of EWS-FLI1. Our results suggest that AHDC1 supports cell growth through EWS-FLI1.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-RBM26 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
Benzonase® Nuclease, Purity > 90%
Sigma-Aldrich
Monoclonal Anti-BRD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1115, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-IRX4 antibody produced in rabbit, IgG fraction of antiserum