Merck
  • Analysis of cardiac myocyte biology in transgenic mice: a protocol for preparation of neonatal mouse cardiac myocyte cultures.
Analysis of cardiac myocyte biology in transgenic mice: a protocol for preparation of neonatal mouse cardiac myocyte cultures.
Methods in molecular biology (Clifton, N.J.) (2010-03-06)
Nigel J Brand, Enrique Lara-Pezzi, Nadia Rosenthal, Paul J R Barton
ABSTRACT
We describe a method of isolating and maintaining primary cultures of mouse neonatal cardiac myocytes (NCM). This is derived from the well-established procedure for making NCM cultures from rat neonates by sequential digestion of rat ventricular myocardial pieces using a collagenase/pancreatin mixture. One-day-old mouse neonates are taken and the heart excised. The great vessels, atria, and top section of the ventricular chambers are cut away and the remaining ventricular myocardium is cut into small cubes (about 1-2 mm(3)). Heart pieces from at least 30 animals are then subjected to short (15-25 min) digestion in a shaking water bath in the presence of collagenase and pancreatin. Cell supernatants are taken and pooled together for a total of five digestion steps. The cells are then plated on gelatinized culture dishes and allowed to attach overnight. Myocyte cultures were inspected microscopically for up to 4 days, revealing that many myocytes beat throughout this period. This protocol may be of use for making primary cardiac myocyte cultures from transgenic mice and for investigating gene transcription and cell signalling.
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Medium 199, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture