Merck
  • Dexmedetomidine Activates Akt, STAT6 and IRF4 Modulating Cytoprotection and Macrophage Anti-Inflammatory Phenotype Against Acute Lung Injury in vivo and in vitro.
Dexmedetomidine Activates Akt, STAT6 and IRF4 Modulating Cytoprotection and Macrophage Anti-Inflammatory Phenotype Against Acute Lung Injury in vivo and in vitro.
Journal of inflammation research (2022-05-04)
Qian Chen, Zhigang Qin, Yibing Sun, Xiangfeng Liu, Aurelie Pac Soo, Enqiang Chang, Qizhe Sun, Bin Yi, Dong-Xin Wang, Hailin Zhao, Daqing Ma, Jianteng Gu
ABSTRACT
This study aims to investigate the cytoprotective and anti-inflammatory effects of an Ī±2-adrenoreceptor (Ī±2-AR) agonist, dexmedetomidine (Dex), on lipopolysaccharides (LPS)-induced acute lung injury and underlying mechanisms with focus on alveolar macrophage polarization modulation. C57BL/6 mice were intraperitoneally injected LPS (10 mg/kg) with or without Dex (25 Āµg/kg) and/or Ī±2-AR antagonist atipamezole (Atip, 500 Āµg/kg). Lung tissues were then analysed to determine injuries. In vitro, human pulmonary epithelial cells (A549) and mice alveolar macrophages (MH-S) were exposed to LPS (10 ng/mL) with or without different concentrations of Dex (0.1-100 nM). Alveolar macrophage polarization, NLRP3 inflammasome activation and inflammatory responses were determined. PTEN/Akt signaling and its downstream transcriptional factors as targets for macrophage polarization were assessed. Dex treatment significantly reduced pro-inflammatory M1 macrophage polarization and NLRP3 inflammasome activation in the lungs relative to the mice treated with LPS. The similar pattern reduction of NLRP3 inflammasome activation by Dex was also found in A549 cells. Atip partly reversed the anti-inflammatory effects of Dex. In cultured alveolar macrophages, Dex reduced LPS-mediated expression of IL-1, -6 and TNF-Ī± receptors while promoting alveolar macrophages differentiation towards a M2 anti-inflammatory phenotype. Additionally, LPS increased Akt signaling activation in a time-dependent manner, which was further activated by Dex via inhibiting phosphatase and tensin homolog (PTEN). The action of Dex on Akt signaling shifted alveolar macrophages from M1 to M2 phenotype through increasing STAT6 and IRF4 transcriptional factors. Dex protected against LPS-induced lung injury and suppressed LPS-induced pulmonary inflammatory responses by attenuating the NLRP3 inflammasome activation and promoting anti-inflammatory M2 macrophage polarization.
Sigma-Aldrich
bpV(pic), ≥95% V basis
Sigma-Aldrich
Dexmedetomidine hydrochloride, ≥98% (HPLC)