Merck
  • Protocol for isolation and proteostatic analysis of sub-populations of spermatogenic cells in mouse.
Protocol for isolation and proteostatic analysis of sub-populations of spermatogenic cells in mouse.
STAR protocols (2022-05-24)
Qianxing Zou, Lele Yang, Huayu Qi
ABSTRACT
Spermatogenesis generates heterologous cell populations which, if not distinguished clearly, often hinder mechanistic and etiological studies. Here, we present a protocol to identify and isolate populations of mouse spermatogenic cells, including spermatogonial stem cells (SSCs), spermatocytes, and haploid spermatids. We also describe absolute quantification of mRNA copy numbers in SSCs. The isolated cells can be used for analyzing nascent protein synthesis and protein degradation, two main events that maintain cellular proteostasis important for healthy and long-term production of male gametes. For complete details on the use and execution of this protocol, please refer to Zou et al. (2021).
Sigma-Aldrich
Micro-dissecting scissors, size 4 1/4 in., straight, sharp point stainless steel
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse