Merck
  • Biodegradation of the sulfonylurea herbicide chlorimuron-ethyl by the strain Pseudomonas sp. LW3.
Biodegradation of the sulfonylurea herbicide chlorimuron-ethyl by the strain Pseudomonas sp. LW3.
FEMS microbiology letters (2009-05-23)
Ji-Ping Ma, Zhe Wang, Peng Lu, Hui-jie Wang, Shinawar Waseem Ali, Shun-Peng Li, Xing Huang
ABSTRACT
The chlorimuron-ethyl-degrading bacterium LW3 was isolated from contaminated soil and identified by 16S rRNA gene sequencing as Pseudomonas sp. When chlorimuron-ethyl was provided as the sole nitrogen source, the degradation efficiency in liquid medium was about 81.0% after 7 days of inoculation with strain LW3. The effects of chlorimuron-ethyl concentration and temperature on biodegradation were examined. Two metabolites of degradation were analyzed by LC/MS. Based on the identified products, strain LW3 seemed to be able to degrade chlorimuron-ethyl by cleavage of the sulfonylurea bridge. The inoculation of strain LW3 to chlorimuron-ethyl-treated soil resulted in a higher degradation rate than in uninoculated soil, regardless of the soil being sterilized or nonsterilized. This microbial culture has great potential for the bioremediation of soil contaminated with chlorimuron-ethyl.
Product Description
Supelco
Chlorimuron ethyl, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chlorimuron ethyl, PESTANAL®, analytical standard