Merck
  • "Shine & Click" Photo-Induced Interfacial Unmasking of Strained Alkynes on Small Water-Soluble Gold Nanoparticles.
"Shine & Click" Photo-Induced Interfacial Unmasking of Strained Alkynes on Small Water-Soluble Gold Nanoparticles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-10-12)
Wilson Luo, Pierangelo Gobbo, Christopher D McNitt, Dewey A Sutton, Vladimir V Popik, Mark S Workentin
ABSTRACT
In this study, we report the design, synthesis, and characterization of small 3 nm water soluble gold nanoparticles (AuNPs) that feature cyclopropenone-masked strained alkyne moieties capable of undergoing interfacial strain-promoted cycloaddition (i-SPAAC) with azides after exposure to UV-A light. A strained alkyne precursor was incorporated onto AuNPs by direct ligand exchange of a thiol-modified cyclopropenone-masked dibenzocyclooctyne (photoDIBO) ligand. These photoDIBO-AuNPs were characterized by 1 H NMR, IR, and UV/Vis spectroscopy, as well as transmission electron microscopy (TEM) and thermogravimetric analysis (TGA), and the extent of modification was quantified. Upon irradiation with UV-A light, photoDIBO-AuNPs underwent efficient and quantitative regeneration of the parent strained alkyne by photochemical decarbonylation to afford DIBO-derivatized AuNPs. DIBO-AuNPs were found to react cleanly and rapidly (k=5.3×10-2  m-1  s-1 ) by an interfacial strain-promoted alkyne-azide cycloadditon (i-SPAAC) with benzyl azide, which served as a simple model system. Furthermore, DIBO-AuNPs were reacted with various azides and a nitrone (interfacial strain-promoted alkyne-nitrone cycloaddition, i-SPANC) to showcase the generality of this approach for the facile modification of AuNP surfaces and their properties. The cyclopropenone-based photo-triggered click chemistry at the interface of water-soluble AuNPs offers exciting opportunities for the atom-by-atom control and assembly of functional materials for applications in materials and biomaterials science as well as in chemical biology.
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 30 nm diameter, Alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 70 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 20 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 60 nm diameter, Alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 50 nm diameter, Alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 10 nm diameter, alkyne functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 80 nm diameter, alkyne functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 90 nm diameter, alkyne functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 100 nm diameter, Alkyne functionalized, 1ml, OD 50, in water