Merck
  • DHHC9-mediated GLUT1 S-palmitoylation promotes glioblastoma glycolysis and tumorigenesis.
DHHC9-mediated GLUT1 S-palmitoylation promotes glioblastoma glycolysis and tumorigenesis.
Nature communications (2021-10-09)
Zhenxing Zhang, Xin Li, Fan Yang, Chao Chen, Ping Liu, Yi Ren, Pengkai Sun, Zixiong Wang, Yongping You, Yi-Xin Zeng, Xinjian Li
ABSTRACT
Glucose transporter GLUT1 is a transmembrane protein responsible for the uptake of glucose into the cells of many tissues through facilitative diffusion. Plasma membrane (PM) localization is essential for glucose uptake by GLUT1. However, the mechanism underlying GLUT1 PM localization remains enigmatic. We find that GLUT1 is palmitoylated at Cys207, and S-palmitoylation is required for maintaining GLUT1 PM localization. Furthermore, we identify DHHC9 as the palmitoyl transferase responsible for this critical posttranslational modification. Knockout of DHHC9 or mutation of GLUT1 Cys207 to serine abrogates palmitoylation and PM distribution of GLUT1, and impairs glycolysis, cell proliferation, and glioblastoma (GBM) tumorigenesis. In addition, DHHC9 expression positively correlates with GLUT1 PM localization in GBM specimens and indicates a poor prognosis in GBM patients. These findings underscore that DHHC9-mediated GLUT1 S-palmitoylation is critical for glucose supply during GBM tumorigenesis.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
APT1 Inhibitor, palmostatin B, The APT1 Inhibitor, palmostatin B controls the biological activity of APT1.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Glucose (GO) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
Anti-phospho GLUT-1 Antibody (Ser226), from rabbit, purified by affinity chromatography