Merck
  • Expression and Purification of Functionally Active Serotonin 5-HT2A Receptor in Insect Cells Using Low-titer Viral Stock.
Expression and Purification of Functionally Active Serotonin 5-HT2A Receptor in Insect Cells Using Low-titer Viral Stock.
Bio-protocol (2021-03-05)
Sukanya Mozumder, Gopa Mahesh, Krishnamoorthi Srinivasan, Jayati Sengupta, Sujoy Mukherjee
ABSTRACT
The serotonin 5-HT2A receptor (5-HT2AR) is a member of the GPCR family that is important for various neurological functions and whose dysregulation causes many mental health disorders. Structural investigations of 5-HT2AR require the production of functionally active receptors expressed from eukaryotic cell cultures. In this protocol, we describe a step-by-step method to express and purify serotonin 5-HT2AR using a baculoviral expression vector system in Sf9 cell cultures, derived from our work with the rat (matching Uniprot ID P14842) and human (matching Uniprot ID P28223) 5-HT2ARs. A unique feature of this method is the utilization of cell culture additives to infect cells at low multiplicity of infection, thereby using several fold less quantity of viral titer compared to prior methods without the additive. This protocol can be tweaked to selectively over-express glycosylated or non-glycosylated forms of the receptor by varying the post-infection harvest times.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
α-Iodoacetamide, α-Iodoacetamide, CAS 144-48-9, is an irreversible inhibitor of several cysteine proteases.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, 0.8 mg/mL, Chemicon®