Merck
  • Prenatal inflammation exposure-programmed hypertension exhibits multi-generational inheritance via disrupting DNA methylome.
Prenatal inflammation exposure-programmed hypertension exhibits multi-generational inheritance via disrupting DNA methylome.
Acta pharmacologica Sinica (2021-10-02)
Xiao Guan, Guo-Rong Dan, Yao Yang, Yan Ji, Wen-Jing Lai, Fang-Jie Wang, Meng Meng, Bang-Hui Mo, Pei Huang, Ting-Ting You, Ya-Fei Deng, Liang Song, Wei Guo, Ping Yi, Jian-Hua Yu, Yuan Gao, Wei-Nian Shou, Bing-Bo Chen, You-Cai Deng, Xiao-Hui Li
ABSTRACT
The multi-generation heredity trait of hypertension in human has been reported, but the molecular mechanisms underlying multi-generational inheritance of hypertension remain obscure. Recent evidence shows that prenatal inflammatory exposure (PIE) results in increased incidence of cardiovascular diseases, including hypertension. In this study we investigated whether and how PIE contributed to multi-generational inheritance of hypertension in rats. PIE was induced in pregnant rats by intraperitoneal injection of LPS or Poly (I:C) either once on gestational day 10.5 (transient stimulation, T) or three times on gestational day 8.5, 10.5, and 12.5 (persistent stimulation, P). Male offspring was chosen to study the paternal inheritance. We showed that PIE, irrespectively induced by LPS or Poly (I:C) stimulation during pregnancy, resulted in multi-generational inheritance of significantly increased blood pressure in rat descendants, and that prenatal LPS exposure led to vascular remodeling and vasoconstrictor dysfunction in both thoracic aorta and superior mesenteric artery of adult F2 offspring. Furthermore, we revealed that PIE resulted in global alteration of DNA methylome in thoracic aorta of F2 offspring. Specifically, PIE led to the DNA hypomethylation of G beta gamma (Gβγ) signaling genes in both the F1 sperm and the F2 thoracic aorta, and activation of PI3K/Akt signaling was implicated in the pathologic changes and dysregulated vascular tone of aortic tissue in F2 LPS-P offspring. Our data demonstrate that PIE reprogrammed DNA methylome of cells from the germline/mature gametes contributes to the development of hypertension in F2 PIE offspring. This study broadens the current knowledge regarding the multi-generation effect of the cumulative early life environmental factors on the development of hypertension.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
USP
Sodium nitroprusside, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, TLR ligand tested