Merck
  • Generation and characterization of CD1d-specific single-domain antibodies with distinct functional features.
Generation and characterization of CD1d-specific single-domain antibodies with distinct functional features.
Immunology (2016-06-18)
Roeland Lameris, Renée C G de Bruin, Paul M P van Bergen En Henegouwen, Henk M Verheul, Sonja Zweegman, Tanja D de Gruijl, Hans J van der Vliet
ABSTRACT
Ligation of the CD1d antigen-presenting molecule by monoclonal antibodies (mAbs) can trigger important biological functions. For therapeutic purposes camelid-derived variable domain of heavy-chain-only antibodies (VHH) have multiple advantages over mAbs because they are small, stable and have low immunogenicity. Here, we generated 21 human CD1d-specific VHH by immunizing Lama glama and subsequent phage display. Two clones induced maturation of dendritic cells, one clone induced early apoptosis in CD1d-expressing B lymphoblasts and multiple myeloma cells, and another clone blocked recognition of glycolipid-loaded CD1d by CD1d-restricted invariant natural killer T (iNKT) cells. In contrast to reported CD1d-specific mAbs, these CD1d-specific VHH have the unique characteristic that they induce specific and well-defined biological effects. This feature, combined with the above-indicated general advantages of VHH, make the CD1d-specific VHH generated here unique and useful tools to exploit both CD1d ligation as well as disruption of CD1d-iNKT interactions in the treatment of cancer or inflammatory disorders.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse