Merck
  • TRPC3 and NALCN channels drive pacemaking in substantia nigra dopaminergic neurons.
TRPC3 and NALCN channels drive pacemaking in substantia nigra dopaminergic neurons.
eLife (2021-08-20)
Ki Bum Um, Suyun Hahn, So Woon Kim, Yoon Je Lee, Lutz Birnbaumer, Hyun Jin Kim, Myoung Kyu Park
ABSTRACT
Midbrain dopamine (DA) neurons are slow pacemakers that maintain extracellular DA levels. During the interspike intervals, subthreshold slow depolarization underlies autonomous pacemaking and determines its rate. However, the ion channels that determine slow depolarization are unknown. Here we show that TRPC3 and NALCN channels together form sustained inward currents responsible for the slow depolarization of nigral DA neurons. Specific TRPC3 channel blockade completely blocked DA neuron pacemaking, but the pacemaking activity in TRPC3 knock-out (KO) mice was perfectly normal, suggesting the presence of compensating ion channels. Blocking NALCN channels abolished pacemaking in both TRPC3 KO and wild-type mice. The NALCN current and mRNA and protein expression are increased in TRPC3 KO mice, indicating that NALCN compensates for TRPC3 currents. In normal conditions, TRPC3 and NALCN contribute equally to slow depolarization. Therefore, we conclude that TRPC3 and NALCN are two major leak channels that drive robust pacemaking in nigral DA neurons.
Sigma-Aldrich
L-703,606 oxalate salt hydrate, solid
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)