Merck
  • SARS-CoV-2 requires cholesterol for viral entry and pathological syncytia formation.
SARS-CoV-2 requires cholesterol for viral entry and pathological syncytia formation.
eLife (2021-04-24)
David W Sanders, Chanelle C Jumper, Paul J Ackerman, Dan Bracha, Anita Donlic, Hahn Kim, Devin Kenney, Ivan Castello-Serrano, Saori Suzuki, Tomokazu Tamura, Alexander H Tavares, Mohsan Saeed, Alex S Holehouse, Alexander Ploss, Ilya Levental, Florian Douam, Robert F Padera, Bruce D Levy, Clifford P Brangwynne
ABSTRACT
Many enveloped viruses induce multinucleated cells (syncytia), reflective of membrane fusion events caused by the same machinery that underlies viral entry. These syncytia are thought to facilitate replication and evasion of the host immune response. Here, we report that co-culture of human cells expressing the receptor ACE2 with cells expressing SARS-CoV-2 spike, results in synapse-like intercellular contacts that initiate cell-cell fusion, producing syncytia resembling those we identify in lungs of COVID-19 patients. To assess the mechanism of spike/ACE2-driven membrane fusion, we developed a microscopy-based, cell-cell fusion assay to screen ~6000 drugs and >30 spike variants. Together with quantitative cell biology approaches, the screen reveals an essential role for biophysical aspects of the membrane, particularly cholesterol-rich regions, in spike-mediated fusion, which extends to replication-competent SARS-CoV-2 isolates. Our findings potentially provide a molecular basis for positive outcomes reported in COVID-19 patients taking statins and suggest new strategies for therapeutics targeting the membrane of SARS-CoV-2 and other fusogenic viruses.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Monensin sodium salt, 90-95% (TLC)
Sigma-Aldrich
Linoleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Dynamin Inhibitor I, Dynasore, The Dynamin Inhibitor I, Dynasore, also referenced under CAS 304448-55-3, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.
Sigma-Aldrich
Nocodazole, Inhibitor of mitosis.
Sigma-Aldrich
Lovastatin, Lovastatin, CAS 75330-75-5, is an anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase.
Sigma-Aldrich
Chlorpromazine, Hydrochloride, Inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase (IC₅₀ = 17 µM).
Sigma-Aldrich
Cholesterol-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Oleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
Bupivacaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Camostat mesylate, ≥98% (HPLC)
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
Sigma-Aldrich
Rostafuroxin, ≥98% (HPLC)
Sigma-Aldrich
Gefitinib, ≥98% (HPLC)
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg