Merck
  • Conformation of Polyethylene Glycol inside Confined Space: Simulation and Experimental Approaches.
Conformation of Polyethylene Glycol inside Confined Space: Simulation and Experimental Approaches.
Nanomaterials (Basel, Switzerland) (2021-01-23)
Tianji Ma, Nicolas Arroyo, Jean Marc Janot, Fabien Picaud, Sebastien Balme
ABSTRACT
The modification of the inner nanopore wall by polymers is currently used to change the specific properties of the nanosystem. Among them, the polyethylene glycol (PEG) is the most used to prevent the fouling and ensure the wettability. However, its properties depend mainly on the chain structure that is very difficult to estimate inside this confined space. Combining experimental and simulation approaches, we provide an insight to the consequence of the PEG presence inside the nanopore on the nanopore properties. We show, in particular, that the cation type in the electrolyte, together with the type of electrolyte (water or urea), is at the origin of the ion transport modification in the nanopore.
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trimethylamine N-oxide dihydrate, purum, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
Trimethylamine N-oxide, 95%