Merck
  • Systematic analysis of purified astrocytes after SCI unveils Zeb2os function during astrogliosis.
Systematic analysis of purified astrocytes after SCI unveils Zeb2os function during astrogliosis.
Cell reports (2021-02-04)
Haichao Wei, Xizi Wu, Yanan You, Raquel Cuevas-Diaz Duran, Yiyan Zheng, K Lakshmi Narayanan, Bo Hai, Xu Li, Neha Tallapragada, Tanuj J Prajapati, Dong H Kim, Benjamin Deneen, Qi-Lin Cao, Jia Qian Wu
ABSTRACT
Spinal cord injury (SCI) is one of the most devastating neural injuries without effective therapeutic solutions. Astrocytes are the predominant component of the scar. Understanding the complex contributions of reactive astrocytes to SCI pathophysiologies is fundamentally important for developing therapeutic strategies. We have studied the molecular changes in the injury environment and the astrocyte-specific responses by astrocyte purification from injured spinal cords from acute to chronic stages. In addition to protein-coding genes, we have systematically analyzed the expression profiles of long non-coding RNAs (lncRNAs) (>200 bp), which are regulatory RNAs that play important roles in the CNS. We have identified a highly conserved lncRNA, Zeb2os, and demonstrated using functional assays that it plays an important role in reactive astrogliosis through the Zeb2os/Zeb2/Stat3 axis. These studies provide valuable insights into the molecular basis of reactive astrogliosis and fill the knowledge gap regarding the function(s) of lncRNAs in astrogliosis and SCI.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
MISSION® esiRNA, targeting human ZEB2
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethanesulfonic acid sodium salt monohydrate, ≥98.0% (T)
Supelco
Serotonin, analytical standard
Sigma-Aldrich
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert
Sigma-Aldrich
MISSION® esiRNA, targeting human AR
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, for molecular biology