Merck
  • Generation of human striatal organoids and cortico-striatal assembloids from human pluripotent stem cells.
Generation of human striatal organoids and cortico-striatal assembloids from human pluripotent stem cells.
Nature biotechnology (2020-12-05)
Yuki Miura, Min-Yin Li, Fikri Birey, Kazuya Ikeda, Omer Revah, Mayuri Vijay Thete, Jin-Young Park, Alyssa Puno, Samuel H Lee, Matthew H Porteus, Sergiu P Pașca
ABSTRACT
Cortico-striatal projections are critical components of forebrain circuitry that regulate motivated behaviors. To enable the study of the human cortico-striatal pathway and how its dysfunction leads to neuropsychiatric disease, we developed a method to convert human pluripotent stem cells into region-specific brain organoids that resemble the developing human striatum and include electrically active medium spiny neurons. We then assembled these organoids with cerebral cortical organoids in three-dimensional cultures to form cortico-striatal assembloids. Using viral tracing and functional assays in intact or sliced assembloids, we show that cortical neurons send axonal projections into striatal organoids and form synaptic connections. Medium spiny neurons mature electrophysiologically following assembly and display calcium activity after optogenetic stimulation of cortical neurons. Moreover, we derive cortico-striatal assembloids from patients with a neurodevelopmental disorder caused by a deletion on chromosome 22q13.3 and capture disease-associated defects in calcium activity, showing that this approach will allow investigation of the development and functional assembly of cortico-striatal connectivity using patient-derived cells.
Sigma-Aldrich
Poly(ethyleneimine) solution, ~50% in H2O
Sigma-Aldrich
SR11237, ≥98% (HPLC)
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%