Merck
Generation and Characterization of Mouse Models for Skeletal Disease.
Methods in molecular biology (Clifton, N.J.) (2020-09-27)
Gabrielle E Foxa, Ye Liu, Lisa M Turner, Alexander G Robling, Tao Yang, Bart O Williams
ABSTRACT
Our laboratories have used genetically engineered mouse models (GEMMs) to assess genetic contributions to skeletal diseases such as osteoporosis and osteoarthritis. Studies on the genetic contributions to OA are often done by assessing how GEMMs respond to surgical methods that induce symptoms modeling OA. Here, we will describe protocols outlining the induction of experimental OA in mice as well as detailed descriptions of methods for analyzing skeletal phenotypes using micro-computerized tomography and skeletal histomorphometry.
Sigma-Aldrich
Alizarin-3-methyliminodiacetic acid
Sigma-Aldrich
Demeclocycline hydrochloride, powder or crystals
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.