Merck
  • 1-alkyl-8-(piperazine-1-sulfonyl)phenylxanthines: development and characterization of adenosine A2B receptor antagonists and a new radioligand with subnanomolar affinity and subtype specificity.
1-alkyl-8-(piperazine-1-sulfonyl)phenylxanthines: development and characterization of adenosine A2B receptor antagonists and a new radioligand with subnanomolar affinity and subtype specificity.
Journal of medicinal chemistry (2009-07-03)
Thomas Borrmann, Sonja Hinz, Daniela C G Bertarelli, Wenjin Li, Nicole C Florin, Anja B Scheiff, Christa E Müller
ABSTRACT
A new series of 1-alkyl-8-(piperazine-1-sulfonyl)phenylxanthines was designed, synthesized, and characterized in radioligand binding and functional assays at A(2B) adenosine receptors. A(2B) antagonists with subnanomolar affinity and high selectivity were discovered. The most potent compounds were 1-ethyl-8-(4-(4-(4-trifluoromethylbenzyl)piperazine-1-sulfonyl)phenyl)xanthine (24, PSB-09120, K(i) (human A(2B)) = 0.157 nM) and 8-(4-(4-(4-chlorobenzyl)piperazine-1-sulfonyl)phenyl)-1-propylxanthine (17, PSB-0788, K(i) (human A(2B)) = 0.393 nM). Moreover, 8-(4-(4-(4-chlorophenyl)piperazine-1-sulfonyl)phenyl)-1-propylxanthine (35, PSB-603) was developed as an A(2B)-specific antagonist exhibiting a K(i) value of 0.553 nM at the human A(2B) receptor and virtually no affinity for the human and rat A(1) and A(2A) and the human A(3) receptors up to a concentration of 10 microM. A tritiated form of the compound was prepared as a new radioligand and characterized in kinetic, saturation, and competition studies. It was shown to be a useful pharmacological tool for the selective labeling of human as well as rodent A(2B) receptors (K(D) human A(2B) 0.403 nM, mouse A(2B) 0.351 nM).
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
PSB-603, ≥98% (HPLC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
2-Chloroadenosine