Merck
  • Inhibition of Rho family GTPases results in increased TNF-alpha production after lipopolysaccharide exposure.
Inhibition of Rho family GTPases results in increased TNF-alpha production after lipopolysaccharide exposure.
Journal of immunology (Baltimore, Md. : 1950) (2003-08-21)
Martha M Monick, Linda S Powers, Noah S Butler, Gary W Hunninghake
ABSTRACT
These studies demonstrate that treatment of macrophages with lovastatin, a cholesterol-lowering drug that blocks farnesylation and geranylgeranylation of target proteins, increases LPS-induced TNF-alpha production. This is reversed by the addition of mevalonate, which bypasses the lovastatin block. Examination of membrane localization of RhoA, Cdc42, Rac1, and Ras demonstrated decreased membrane localization of the geranylgeranylated Rho family members (RhoA, Cdc42, and Rac1) with no change in the membrane localization of farnesylated Ras. LPS-induced TNF-alpha production in the presence of the Rho family-specific blocker (toxin B from Clostridium difficile) was significantly enhanced consistent with the lovastatin data. One intracellular signaling pathway that is required for TNF-alpha production by LPS is the extracellular signal-regulated kinase (ERK). Significantly, we found prolonged ERK activation after LPS stimulation of lovastatin-treated macrophages. When we inhibited ERK, we blocked the lovastatin-induced increase in TNF-alpha production. As a composite, these studies demonstrate a negative role for one or more Rho family GTPases in LPS-induced TNF-alpha production.
Product Description
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
Sigma-Aldrich
Lovastatin, Lovastatin, CAS 75330-75-5, is an anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase.
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
Anti-Ras Antibody, clone RAS10, clone RAS10, Upstate®, from mouse