Merck
  • CircSMYD4 regulates proliferation, migration and apoptosis of hepatocellular carcinoma cells by sponging miR-584-5p.
CircSMYD4 regulates proliferation, migration and apoptosis of hepatocellular carcinoma cells by sponging miR-584-5p.
Cancer cell international (2020-12-10)
Yanhe Zhang, Hui Wang, Chao Li, Linlin Gao, Yayun Zheng, Wenjuan Chang, Chao Lu, Xiaoguang Zhao
ABSTRACT
There is evidence that circSMYD4 is differentially expressed in hepatocellular carcinoma (HCC), but its mechanism of action remains unclear. Therefore, this study aimed to explore the role of circSMYD4 in the occurrence and development of HCC and its specific molecular mechanism. The expressions of related genes and proteins in the development of HCC were detected by real-time quantitative-PCR and Western blot. HCC cells treated with RNase R and Actinomycin D were used to examine the stability of circSMYD4. Bioinformatics analysis, RNA pull-down assay, luciferase assay and Spearman correlation analysis were performed to evaluate the interaction between circSMYD4 and miRNA. Cell Counting Kit-8, clone formation assay, wound healing assay, Transwell, flow cytometry, nude tumor formation experiment, and immunohistochemistry were employed to analyze the function of circSMYD4 in HCC. A rescue experiment was conducted to analyze the effect of miR-584-5p on the physiological functions of cells. CircSMYD4 was down-regulated in HCC tissues and cells, and was not easily affected by RNase R and Actinomycin D. The abundances of circSMYD4 and SMYD4 in the cytoplasm were significantly higher than in the nucleus. Up-regulation of circSMYD4 inhibited the proliferation, invasion and migration and promoted the apoptosis of HCC cells in vitro, while it inhibited tumor growth, promoted apoptosis-related proteins, and suppressed alpha-fetoprotein (AFP) levels in vivo. CircSMYD4 could be used as a miRNA sponge to target miR-584-5p. In addition, miR-584-5p overexpression partially reversed the regulatory effect of circSMYD4 on HCC. CircSMYD4 prevents the development of HCC through regulating multiple signaling pathways such as metastasis and apoptosis by sponging miR-584-5p.
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Annexin V-FITC Apoptosis Detection Kit
Corning® HTS Transwell®-24 well permeable supports, HTS Transwell-24 units w/ 3.0 μm pore polycarbonate membrane and 6.5 mm inserts, TC-treated, sterile, 2/cs
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder