Merck
  • Pilocarpine-induced seizures associate with modifications of LSD1/CoREST/HDAC1/2 epigenetic complex and repressive chromatin in mice hippocampus.
Pilocarpine-induced seizures associate with modifications of LSD1/CoREST/HDAC1/2 epigenetic complex and repressive chromatin in mice hippocampus.
Biochemistry and biophysics reports (2021-01-12)
Verónica Noches, Carlos Rivera, Marcela P González, Gianluca Merello, Montserrat Olivares-Costa, María Estela Andrés
ABSTRACT
Epilepsy is a neurological disorder of genetic or environmental origin characterized by recurrent spontaneous seizures. A rodent model of temporal lobe epilepsy is induced by a single administration of pilocarpine, a non-selective cholinergic muscarinic receptor agonist. The molecular changes associated with pilocarpine-induced seizures are still poorly described. Epigenetic multiprotein complexes that regulate gene expression by changing the structure of chromatin impose transcriptional memories. Among the epigenetic enzymes relevant to the epileptogenic process is lysine-specific demethylase 1 (LSD1, KDM1A), which regulates the expression of genes that control neuronal excitability. LSD1 forms complexes with the CoREST family of transcriptional corepressors, which are molecular bridges that bring HDAC1/2 and LSD1 enzymes to deacetylate and demethylate the tail of nucleosomal histone H3. To test the hypothesis that LSD1-complexes are involved in initial modifications associated with pilocarpine-induced epilepsy, we studied the expression of main components of LSD1-complexes and the associated epigenetic marks on isolated neurons and the hippocampus of pilocarpine-treated mice. Using a single injection of 300 mg/kg of pilocarpine and after 24 h, we found that protein levels of LSD1, CoREST2, and HDAC1/2 increased, while CoREST1 decreased in the hippocampus. In addition, we observed increased histone H3 lysine 9 di- and trimethylation (H3K9me2/3) and decreased histone H3 lysine 4 di and trimethylation (H3K4me2/3). Similar findings were observed in cultured hippocampal neurons and HT-22 hippocampal cell line treated with pilocarpine. In conclusion, our data show that muscarinic receptor activation by pilocarpine induces a global repressive state of chromatin and prevalence of LSD1-CoREST2 epigenetic complexes, modifications that could underlie the pathophysiological processes leading to epilepsy.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
HT-22 Mouse Hippocampal Neuronal Cell Line, HT-22 mouse neuronal cell line is a valuable cell model for studies of glutamate-induced toxicity in neuronal cells.
Sigma-Aldrich
Anti-RCOR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution