Merck
  • Requisite Chromatin Remodeling for Myeloid and Erythroid Lineage Differentiation from Erythromyeloid Progenitors.
Requisite Chromatin Remodeling for Myeloid and Erythroid Lineage Differentiation from Erythromyeloid Progenitors.
Cell reports (2020-11-19)
Jun Wu, Karen Krchma, Hyung Joo Lee, Sairam Prabhakar, Xiaoli Wang, Haiyong Zhao, Xiaoyun Xing, Rho H Seong, Daved H Fremont, Maxim N Artyomov, Ting Wang, Kyunghee Choi
ABSTRACT
The mammalian SWitch/Sucrose Non-Fermentable (SWI/SNF) chromatin-remodeling BAF (BRG1/BRM-associated factor) complex plays an essential role in developmental and pathological processes. We show that the deletion of Baf155, which encodes a subunit of the BAF complex, in the Tie2(+) lineage (Baf155 (CKO) leads to defects in yolk sac myeloid and definitive erythroid (EryD) lineage differentiation from erythromyeloid progenitors (EMPs). The chromatin of myeloid gene loci in Baf155 CKO EMPs is mostly inaccessible and enriched mainly by the ETS binding motif. BAF155 interacts with PU.1 and is recruited to PU.1 target gene loci together with p300 and KDM6a. Treatment of Baf155 CKO embryos with GSK126, an H3K27me2/3 methyltransferase EZH2 inhibitor, rescues myeloid lineage gene expression. This study uncovers indispensable BAF-mediated chromatin remodeling of myeloid gene loci at the EMP stage. Future studies exploiting epigenetics in the generation and application of EMP derivatives for tissue repair, regeneration, and disease are warranted.
Product Description
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Millipore
EZview Red Anti-HA Affinity Gel
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
MISSION® pLKO.1-puro-UbC-TurboGFP Positive Control Plasmid DNA, Contains a gene encoding TurboGFP, under the control of the UbC promoter
Sigma-Aldrich
MISSION® esiRNA, targeting human NRGN
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Smarcc1
Sigma-Aldrich
MISSION® esiRNA, targeting EGFP
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Supelco
N-(Propionyloxy)succinimide, for LC-MS derivatization, LiChropur, ≥95% (HPLC)