Merck
  • Nrf2-dependent and -independent effects of tBHQ in activated murine B cells.
Nrf2-dependent and -independent effects of tBHQ in activated murine B cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-07-24)
Jenna K Bursley, Cheryl E Rockwell, Jenna K Bursley, Cheryl E Rockwell
ABSTRACT
Nrf2 is a transcription factor that regulates cytoprotective cellular responses to oxidative and electrophilic stress. Nrf2 is potently activated by the synthetic food additive, tert-butylhydroquinone (tBHQ), which is widely used as a preservative in oils and processed foods. Previously published studies have established that tBHQ has numerous effects on T cell function. The purpose of this study was to determine the effect of tBHQ on B cell function and the role of Nrf2 in these effects. Specifically, we investigated T cell-independent B cell activation, differentiation, and IgM antibody production. Murine wild-type and Nrf2-null splenocytes were isolated, treated with tBHQ (0.25-2.5 μm), and activated by lipopolysaccharide (LPS), a T cell-independent B cell activator. Our findings indicate that tBHQ significantly enhanced IgM production in activated wild-type, but not Nrf2-null, B cells, suggesting this effect is Nrf2-dependent. In contrast, tBHQ significantly decreased the induction of CD69, CD25, CD22, and CD138 in both wild-type and Nrf2-null splenocytes. These findings indicate that the tBHQ-mediated increase in IgM is Nrf2-dependent, whereas the inhibition of CD69, CD25, CD22 and CD138 is Nrf2-independent. Overall, this study demonstrates that in addition to its effects on T cells, tBHQ also has potent effects on T cell-independent B cell function.
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IgM, Kappa from murine myeloma, clone TEPC 183, purified immunoglobulin, buffered aqueous solution