Merck
  • The BRD7-P53-SLC25A28 axis regulates ferroptosis in hepatic stellate cells.
The BRD7-P53-SLC25A28 axis regulates ferroptosis in hepatic stellate cells.
Redox biology (2020-08-31)
Zili Zhang, Mei Guo, Min Shen, Desong Kong, Feng Zhang, Jiangjuan Shao, Shanzhong Tan, Shijun Wang, Anping Chen, Peng Cao, Shizhong Zheng
ABSTRACT
Ferroptosis is a recently discovered form of programmed cell death, but its regulatory mechanisms are not fully understood. In the current study, we reported that the BRD7-P53-SLC25A28 axis played a crucial role in regulating ferroptosis in hepatic stellate cells (HSCs). Upon exposure to ferroptosis inducers, bromodomain-containing protein 7 (BRD7) protein expression was remarkably increased through the inhibition of the ubiquitin-proteasome pathway. CRISPR/Cas9-mediated BRD7 knockout conferred resistance to HSC ferroptosis, whereas specific BRD7 plasmid-mediated BRD7 overexpression facilitated HSC ferroptosis. Interestingly, the elevated BRD7 expression exhibited to promote p53 mitochondrial translocation via direct binding with p53 N-terminal transactivation domain (TAD), which may be the underlying mechanisms for BRD7-enhanced HSC ferroptosis. Site-directed mutations of serine 392 completely blocked the binding of BRD7 to p53, and, in turn, prevented p53 mitochondrial translocation and HSC ferroptosis. Importantly, mitochondrial p53 interacted with solute carrier family 25 member 28 (SLC25A28) to form complex and enhanced the activity of SLC25A28, which could lead to the abnormal accumulation of redox-active iron and hyperfunction of electron transfer chain (ETC). SLC25A28 knockdown impaired BRD7-or p53-mediated ferroptotic events. In mice, erastin treatment ameliorated pathological damage of liver fibrosis through inducing HSC ferroptosis. HSC-specific blockade of BRD7-P53-SLC25A28 axis could abrogate erastin-induced HSC ferroptosis. Of note, we analyzed the effect of sorafenib on HSC ferroptosis in advanced fibrotic patients with hepatocellular carcinoma receiving sorafenib monotherapy. Attractively, BRD7 upregulation, p53 mitochondrial translocation, combination of SLC25A28 and p53, and ferroptosis induction occurred in primary human HSCs. Overall, these findings reveal novel signal transduction and regulatory mechanism of ferroptosis, and also suggest BRD7-P53-SLC25A28 axis as potential targets for liver fibrosis.
Sigma-Aldrich
Glutathione Assay Kit, sufficient for 700 assays
Sigma-Aldrich
MISSION® esiRNA, targeting human BRD7
Sigma-Aldrich
Universal Antibody Dilution Buffer, Ready-To-Use, with BSA
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Hanks′ Balanced Salt Solution 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Immobilon®-P Polyvinylidene difluoride membranes, size 26.5 cm × 375 cm
Sigma-Aldrich
Mammalian Cell Lysis Kit
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins–FITC antibody produced in mouse, clone RG-16, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei PURE Prep, sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Corning® cell strainer, pore size 70 μm, white, sterile, pkg of (individually wrapped), pack of 50 ea, Corning 431751
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Corning® tissue-culture treated culture dishes, D × H 150 mm × 25 mm
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC25A28
Sigma-Aldrich
Diethyl butylmalonate, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)