Merck
  • Prostate Cancer Proliferation Is Affected by the Subcellular Localization of MCT2 and Accompanied by Significant Peroxisomal Alterations.
Prostate Cancer Proliferation Is Affected by the Subcellular Localization of MCT2 and Accompanied by Significant Peroxisomal Alterations.
Cancers (2020-10-31)
Isabel Valença, Ana Rita Ferreira, Marcelo Correia, Sandra Kühl, Carlo van Roermund, Hans R Waterham, Valdemar Máximo, Markus Islinger, Daniela Ribeiro
ABSTRACT
Reprogramming of lipid metabolism directly contributes to malignant transformation and progression. The increased uptake of circulating lipids, the transfer of fatty acids from stromal adipocytes to cancer cells, the de novo fatty acid synthesis, and the fatty acid oxidation support the central role of lipids in many cancers, including prostate cancer (PCa). Fatty acid β-oxidation is the dominant bioenergetic pathway in PCa and recent evidence suggests that PCa takes advantage of the peroxisome transport machinery to target monocarboxylate transporter 2 (MCT2) to peroxisomes in order to increase β-oxidation rates and maintain the redox balance. Here we show evidence suggesting that PCa streamlines peroxisome metabolism by upregulating distinct pathways involved in lipid metabolism. Moreover, we show that MCT2 is required for PCa cell proliferation and, importantly, that its specific localization at the peroxisomal membranes is essential for this role. Our results highlight the importance of peroxisomes in PCa development and uncover different cellular mechanisms that may be further explored as possible targets for PCa therapy.
Sigma-Aldrich
Anti-ACBD5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-PEX19 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-ACOX3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PMP70 antibody, Mouse monoclonal, clone 70-18, purified from hybridoma cell culture