Merck
  • Intranasal delivery of cationic PLGA nano/microparticles-loaded FMDV DNA vaccine encoding IL-6 elicited protective immunity against FMDV challenge.
Intranasal delivery of cationic PLGA nano/microparticles-loaded FMDV DNA vaccine encoding IL-6 elicited protective immunity against FMDV challenge.
PloS one (2011-11-24)
Gang Wang, Li Pan, Yongguang Zhang, Yonglu Wang, Zhongwang Zhang, Jianliang Lü, Peng Zhou, Yuzhen Fang, Shoutian Jiang
ABSTRACT
Mucosal vaccination has been demonstrated to be an effective means of eliciting protective immunity against aerosol infections of foot and mouth disease virus (FMDV) and various approaches have been used to improve mucosal response to this pathogen. In this study, cationic PLGA (poly(lactide-co-glycolide)) nano/microparticles were used as an intranasal delivery vehicle as a means administering FMDV DNA vaccine encoding the FMDV capsid protein and the bovine IL-6 gene as a means of enhancing mucosal and systemic immune responses in animals. Three eukaryotic expression plasmids with or without bovine IL-6 gene (pc-P12A3C, pc-IL2AP12A3C and pc-P12AIL3C) were generated. The two latter plasmids were designed with the IL-6 gene located either before or between the P12A and 3C genes, respectively, as a means of determining if the location of the IL-6 gene affected capsid assembly and the subsequent immune response. Guinea pigs and rats were intranasally vaccinated with the respective chitosan-coated PLGA nano/microparticles-loaded FMDV DNA vaccine formulations. Animals immunized with pc-P12AIL3C (followed by animals vaccinated with pc-P12A3C and pc-IL2AP12A3C) developed the highest levels of antigen-specific serum IgG and IgA antibody responses and the highest levels of sIgA (secretory IgA) present in mucosal tissues. However, the highest levels of neutralizing antibodies were generated in pc-IL2AP12A3C-immunized animals (followed by pc-P12AIL3C- and then in pc-P12A3C-immunized animals). pc-IL2AP12A3C-immunized animals also developed stronger cell mediated immune responses (followed by pc-P12AIL3C- and pc-P12A3C-immunized animals) as evidenced by antigen-specific T-cell proliferation and expression levels of IFN-γ by both CD4+ and CD8+ splenic T cells. The percentage of animals protected against FMDV challenge following immunizations with pc-IL2AP12A3C, pc-P12AIL3C or pc-P12A3C were 3/5, 1/5 and 0/5, respectively. These data suggested that intranasal delivery of cationic PLGA nano/microparticles loaded with various FMDV DNA vaccine formulations encoding IL-6 as a molecular adjuvant enhanced protective immunity against FMDV, particularly pc-IL2AP12A3C with IL-6 gene located before P12A3C gene.
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Saponin, used as non-ionic surfactant