Merck
64-Channel Carbon Fiber Electrode Arrays for Chronic Electrophysiology.
Scientific reports (2020-03-04)
Grigori Guitchounts, David Cox
ABSTRACT
A chief goal in neuroscience is to understand how neuronal activity relates to behavior, perception, and cognition. However, monitoring neuronal activity over long periods of time is technically challenging, and limited, in part, by the invasive nature of recording tools. While electrodes allow for recording in freely-behaving animals, they tend to be bulky and stiff, causing damage to the tissue they are implanted in. One solution to this invasiveness problem may be probes that are small enough to fly under the immune system's radar. Carbon fiber (CF) electrodes are thinner and more flexible than typical metal or silicon electrodes, but the arrays described in previous reports had low channel counts and required time-consuming manual assembly. Here we report the design of an expanded-channel-count carbon fiber electrode array (CFEA) as well as a method for fast preparation of the recording sites using acid etching and electroplating with PEDOT-TFB, and demonstrate the ability of the 64-channel CFEA to record from rat visual cortex. We include designs for interfacing the system with micro-drives or flex-PCB cables for recording from multiple brain regions, as well as a facilitated method for coating CFs with the insulator Parylene-C. High-channel-count CFEAs may thus be an alternative to traditional microwire-based electrodes and a practical tool for exploring the neural code.
Sigma-Aldrich
3,4-Ethylenedioxythiophene, 97%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Tetrabutylammonium tetrafluoroborate, for electrochemical analysis, ≥99.0%