Merck
  • Sexual dimorphism in the meiotic requirement for PRDM9: A mammalian evolutionary safeguard.
Sexual dimorphism in the meiotic requirement for PRDM9: A mammalian evolutionary safeguard.
Science advances (2020-10-25)
Natalie R Powers, Beth L Dumont, Chihiro Emori, Raman Akinyanju Lawal, Catherine Brunton, Kenneth Paigen, Mary Ann Handel, Ewelina Bolcun-Filas, Petko M Petkov, Tanmoy Bhattacharyya
ABSTRACT
In many mammals, genomic sites for recombination are determined by the histone methyltransferase PRMD9. Some mouse strains lacking PRDM9 are infertile, but instances of fertility or semifertility in the absence of PRDM9 have been reported in mice, canines, and a human female. Such findings raise the question of how the loss of PRDM9 is circumvented to maintain fertility. We show that genetic background and sex-specific modifiers can obviate the requirement for PRDM9 in mice. Specifically, the meiotic DNA damage checkpoint protein CHK2 acts as a modifier allowing female-specific fertility in the absence of PRDM9. We also report that, in the absence of PRDM9, a PRDM9-independent recombination system is compatible with female meiosis and fertility, suggesting sex-specific regulation of meiotic recombination, a finding with implications for speciation.
Sigma-Aldrich
Donkey Anti-Guinea Pig IgG Antibody, HRP conjugate, Species Adsorbed, 1 mg/mL (Reconstituted), Chemicon®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Goat Anti-Guinea Pig IgG Antibody, Cy3 conjugate, Chemicon®, from goat
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-phospho-H2A.X (Ser139) Antibody, Upstate®, from rabbit