Merck
  • nNOS-expressing interneurons control basal and behaviorally evoked arterial dilation in somatosensory cortex of mice.
nNOS-expressing interneurons control basal and behaviorally evoked arterial dilation in somatosensory cortex of mice.
eLife (2020-10-06)
Christina T Echagarruga, Kyle W Gheres, Jordan N Norwood, Patrick J Drew
ABSTRACT
Cortical neural activity is coupled to local arterial diameter and blood flow. However, which neurons control the dynamics of cerebral arteries is not well understood. We dissected the cellular mechanisms controlling the basal diameter and evoked dilation in cortical arteries in awake, head-fixed mice. Locomotion drove robust arterial dilation, increases in gamma band power in the local field potential (LFP), and increases calcium signals in pyramidal and neuronal nitric oxide synthase (nNOS)-expressing neurons. Chemogenetic or pharmocological modulation of overall neural activity up or down caused corresponding increases or decreases in basal arterial diameter. Modulation of pyramidal neuron activity alone had little effect on basal or evoked arterial dilation, despite pronounced changes in the LFP. Modulation of the activity of nNOS-expressing neurons drove changes in the basal and evoked arterial diameter without corresponding changes in population neural activity.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 70,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
Muscimol, powder
Sigma-Aldrich
Clozapine N-oxide
Supelco
Mesosulfuron-methyl, PESTANAL®, analytical standard
Sigma-Aldrich
ML133 hydrochloride, ≥95% (HPLC)