Merck
  • Exploring the Enzymatic and Antibacterial Activities of Novel Mycobacteriophage Lysin B Enzymes.
Exploring the Enzymatic and Antibacterial Activities of Novel Mycobacteriophage Lysin B Enzymes.
International journal of molecular sciences (2020-05-06)
Adel Abouhmad, Ahmed H Korany, Carl Grey, Tarek Dishisha, Rajni Hatti-Kaul
ABSTRACT
Mycobacteriophages possess different sets of lytic enzymes for disruption of the complex cell envelope of the mycobacteria host cells and release of the viral progeny. Lysin B (LysB) enzymes are mycolylarabinogalactan esterases that cleave the ester bond between the arabinogalactan and mycolic acids in the mycolylarabinogalactan-peptidoglycan (mAGP) complex in the cell envelope of mycobacteria. In the present study, four LysB enzymes were produced recombinantly and characterized with respect to their enzymatic and antibacterial activities. Examination of the kinetic parameters for the hydrolysis of para-nitrophenyl ester substrates, shows LysB-His6 enzymes to be active against a range of substrates (C4-C16), with a catalytic preference towards p-nitrophenyl laurate (C12). With p-nitrophenyl butyrate as substrate, LysB-His6 enzymes showed highest activity at 37 °C. LysB-His6 enzymes also hydrolyzed different Tween substrates with highest activity against Tween 20 and 80. Metal ions like Ca2+ and Mn2+ enhanced the enzymatic activity of LysB-His6 enzymes, while transition metal ions like Zn2+ and Cu2+ inhibited the enzymatic activity. The mycolylarabinogalactan esterase activity of LysB-His6 enzymes against mAGP complex was confirmed by LC-MS. LysB-His6 enzymes showed marginal antibacterial activity when tested alone against Mycobacterium smegmatis, however a synergetic activity was noticed when combined with outer membrane permealizers. These results confirm that LysB enzymes are lipolytic enzymes with potential application as antimycobacterials.
Sigma-Aldrich
4-Nitrophenyl octanoate, ≥90.0% (GC)
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate
Sigma-Aldrich
4-Nitrophenyl stearate, lipase substrate
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%
Sigma-Aldrich
4-Nitrophenyl myristate, ≥95.0% (HPLC)