Merck
  • Phosphorylation of eukaryotic initiation factor-2α (eIF2α) in autophagy.
Phosphorylation of eukaryotic initiation factor-2α (eIF2α) in autophagy.
Cell death & disease (2020-06-10)
Juliette Humeau, Marion Leduc, Giulia Cerrato, Friedemann Loos, Oliver Kepp, Guido Kroemer
ABSTRACT
The integrated stress response is characterized by the phosphorylation of eukaryotic initiation factor-2α (eIF2α) on serine 51 by one out of four specific kinases (EIF2AK1 to 4). Here we provide three series of evidence suggesting that macroautophagy (to which we refer to as autophagy) induced by a variety of distinct pharmacological agents generally requires this phosphorylation event. First, the induction of autophagic puncta by various distinct compounds was accompanied by eIF2α phosphorylation on serine 51. Second, the modulation of autophagy by >30 chemically unrelated agents was partially inhibited in cells expressing a non-phosphorylable (S51A) mutant of eIF2α or lacking all four eIF2α kinases, although distinct kinases were involved in the response to different autophagy inducers. Third, inhibition of eIF2α phosphatases was sufficient to stimulate autophagy. In synthesis, it appears that eIF2α phosphorylation is a central event for the stimulation of autophagy.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate, ≥98.0%
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5% (titration)
Sigma-Aldrich
1-Cyano-4-dimethylaminopyridinium tetrafluoroborate, organic cyanylating reagent
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Vinblastine sulfate salt, ≥97% (HPLC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Anacardic acid
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
UK-5099, ≥98% (HPLC)
Sigma-Aldrich
Nelfinavir mesylate hydrate, ≥98% (HPLC)
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Crizotinib, ≥98% (HPLC)
Sigma-Aldrich
Spermidine-(butyl-13C4) trihydrochloride, 99 atom % 13C, 95% (CP)
Sigma-Aldrich
Quercetin hydrate, ≥95%
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Guanabenz acetate salt, powder
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
LentiBrite RFP-LC3 Lentiviral Biosensor
Sigma-Aldrich
Sephin1, ≥95% (HPLC)
Sigma-Aldrich
Salubrinal, ≥98% (HPLC)