Merck
  • A critical role of PRDM14 in human primordial germ cell fate revealed by inducible degrons.
A critical role of PRDM14 in human primordial germ cell fate revealed by inducible degrons.
Nature communications (2020-03-11)
Anastasiya Sybirna, Walfred W C Tang, Merrick Pierson Smela, Sabine Dietmann, Wolfram H Gruhn, Ran Brosh, M Azim Surani
ABSTRACT
PRDM14 is a crucial regulator of mouse primordial germ cells (mPGCs), epigenetic reprogramming and pluripotency, but its role in the evolutionarily divergent regulatory network of human PGCs (hPGCs) remains unclear. Besides, a previous knockdown study indicated that PRDM14 might be dispensable for human germ cell fate. Here, we decided to use inducible degrons for a more rapid and comprehensive PRDM14 depletion. We show that PRDM14 loss results in significantly reduced specification efficiency and an aberrant transcriptome of hPGC-like cells (hPGCLCs) obtained in vitro from human embryonic stem cells (hESCs). Chromatin immunoprecipitation and transcriptomic analyses suggest that PRDM14 cooperates with TFAP2C and BLIMP1 to upregulate germ cell and pluripotency genes, while repressing WNT signalling and somatic markers. Notably, PRDM14 targets are not conserved between mouse and human, emphasising the divergent molecular mechanisms of PGC specification. The effectiveness of degrons for acute protein depletion is widely applicable in various developmental contexts.
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-PRDM14 Antibody, from rabbit
Sigma-Aldrich
Allyl methyl sulfone, 96%