Merck
  • α-Linolenic Acid-Enriched Cold-Pressed Perilla Oil Suppress High-Fat Diet-Induced Hepatic Steatosis through Amelioration of the ER Stress-Mediated Autophagy.
α-Linolenic Acid-Enriched Cold-Pressed Perilla Oil Suppress High-Fat Diet-Induced Hepatic Steatosis through Amelioration of the ER Stress-Mediated Autophagy.
Molecules (Basel, Switzerland) (2020-06-12)
Su Ji Bae, Ji Eun Kim, Hyeon Jun Choi, Yun Ju Choi, Su Jin Lee, Jeong Eun Gong, Sungbaek Seo, Seung Yun Yang, Beum-Soo An, Hee Seob Lee, Dong Seob Kim, Chung Yeoul Lee, Dae Youn Hwang
ABSTRACT
Perilla oil has been considered to have excellent potential for treating various diseases due to its contents of beneficial fatty acids, such as α-linolenic acid, oleic acid and linoleic acid. The therapeutic effects and molecular mechanism of an α-linolenic acid-enriched cold-pressed perilla oil (LEP) on hepatic steatosis of an obesity model were investigated by analyzing alterations in fat accumulation and endoplasmic reticulum (ER) stress-mediated autophagy, in high-fat diet (HFD)-induced obesity C57BL/6N mice treated with LEP for 16 weeks. Although no significant alterations were detected in body weight and most organ weights, the liver weight and accumulation of lipid droplets in the liver section were significantly lower in HFD + LEP treated group as compared to the HFD + Vehicle treated group. Reduced mRNA expression levels of adipogenesis and lipogenesis regulating factors, including the peroxisome proliferator-activated receptor (PPAR)γ, CCAAT/enhancer-binding protein (C/EBP)α, fatty acid synthase (FAS), and adipocyte fatty acid-binding protein 2 (aP2) were observed after LEP treatment for 16 weeks, while the levels of lipolysis were remarkably increased in the same group. Moreover, the LEP-treated groups showed suppression of ER stress-regulating factors, such as the C/EBP homologous protein (CHOP), eukaryotic translation initiation factor 2α (eIF2α), inositol-requiring protein 1 (IRE1)α, and Jun-N-terminal kinase (JNK) during anti-hepatic steatosis effects. The expression level of the microtubule-associated protein 1A/1B-light chain 3 (LC3) protein and phosphatidylinositol-3-kinase (PI3K)/AKT/ mammalian target of rapamycin (mTOR) pathway for the autophagy response showed a significant decrease in the HFD+LEP-treated group. Furthermore, ER stress-mediated autophagy was accompanied with enhanced phosphorylation of extracellular signal-regulated kinase (ERK), JNK, and p38 protein in the mitogen-activated protein (MAP) kinase signaling pathway. Taken together, the results of the present study indicate that treatment with LEP inhibits hepatic steatosis in the HFD-induced obese model through regulation of adipogenesis and lipolysis. We believe our results are the first to show that the anti-hepatic steatosis activity of α-linolenic acid from cold-pressed perilla oil might be tightly correlated with the amelioration of ER stress-mediated autophagy.
Sigma-Aldrich
Eosin Y Solution, Alcoholic, with Phloxine
Sigma-Aldrich
Mayer′s Hematoxylin Solution