Merck
  • Sacrificial Fibers Improve Matrix Distribution and Micromechanical Properties in a Tissue-Engineered Intervertebral Disc.
Sacrificial Fibers Improve Matrix Distribution and Micromechanical Properties in a Tissue-Engineered Intervertebral Disc.
Acta biomaterialia (2020-05-25)
Beth G Ashinsky, Sarah E Gullbrand, Edward D Bonnevie, Chao Wang, Dong Hwa Kim, Lin Han, Robert L Mauck, Harvey E Smith
ABSTRACT
Tissue-engineered replacement discs are an area of intense investigation for the treatment of end-stage intervertebral disc (IVD) degeneration. These living implants can integrate into the IVD space and recapitulate native motion segment function. We recently developed a multiphasic tissue-engineered disc-like angle-ply structure (DAPS) that models the micro-architectural and functional features of native tissue. While these implants resulted in functional restoration of the motion segment in rat and caprine models, we also noted deficiencies in cell infiltration and homogeneity of matrix deposition in the electrospun poly(ε-caprolactone) outer region (annulus fibrosus, AF) of the DAPS. To address this limitation, here, we incorporated a sacrificial water-soluble polymer, polyethylene oxide (PEO), as a second fiber fraction within the AF region to increase porosity of the implant. Maturation of these PEO-modified DAPS were evaluated after 5 and 10 weeks of in vitro culture in terms of AF biochemical content, MRI T2 values, overall construct mechanical properties, AF micromechanical properties and cell and matrix distribution. To assess the performance of the PEO-modified DAPS in vivo, precultured constructs were implanted into the rat caudal IVD space for 10 weeks. Results showed that matrix distribution was more homogenous in PCL/PEO DAPS, as evidenced by more robust histological staining, organized collagen deposition and micromechanical properties, compared to standard PCL-only DAPS in vitro. Cell and matrix infiltration were also improved in vivo, but no differences in macromechanical properties and a trend towards improved micromechanical properties were observed. These findings demonstrate that the inclusion of a sacrificial PEO fiber fraction in the DAPS AF region improves cellular colonization, matrix elaboration, and in vitro and in vivo function of an engineered IVD implant. STATEMENT OF SIGNIFICANCE: This work establishes a method for improving cell infiltration and matrix distribution within tissue-engineered dense fibrous scaffolds for intervertebral disc replacement. Tissue-engineered whole disc replacements are an attractive alternative to the current gold standard (mechanical disc arthroplasty or vertebral fusion) for the clinical treatment of patients with advanced disc degeneration.
Sigma-Aldrich
Eosin Y Solution, Alcoholic
Sigma-Aldrich
Scott′s Tap Water Substitute Concentrate, 10 ×
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Hematoxylin Solution, Gill No. 2