Merck
  • Cathepsin C is a novel mediator of podocyte and renal injury induced by hyperglycemia.
Cathepsin C is a novel mediator of podocyte and renal injury induced by hyperglycemia.
Biochimica et biophysica acta. Molecular cell research (2020-04-18)
Irena Audzeyenka, Patrycja Rachubik, Dorota Rogacka, Marlena Typiak, Tomasz Kulesza, Stefan Angielski, Michał Rychłowski, Magdalena Wysocka, Natalia Gruba, Adam Lesner, Moin A Saleem, Agnieszka Piwkowska
ABSTRACT
A growing body of evidence suggests a role of proteolytic enzymes in the development of diabetic nephropathy. Cathepsin C (CatC) is a well-known regulator of inflammatory responses, but its involvement in podocyte and renal injury remains obscure. We used Zucker rats, a genetic model of metabolic syndrome and insulin resistance, to determine the presence, quantity, and activity of CatC in the urine. In addition to the animal study, we used two cellular models, immortalized human podocytes and primary rat podocytes, to determine mRNA and protein expression levels via RT-PCR, Western blot, and confocal microscopy, and to evaluate CatC activity. The role of CatC was analyzed in CatC-depleted podocytes using siRNA and glycolytic flux parameters were obtained from extracellular acidification rate (ECAR) measurements. In functional analyses, podocyte and glomerular permeability to albumin was determined. We found that podocytes express and secrete CatC, and a hyperglycemic environment increases CatC levels and activity. Both high glucose and non-specific activator of CatC phorbol 12-myristate 13-acetate (PMA) diminished nephrin, cofilin, and GLUT4 levels and induced cytoskeletal rearrangements, increasing albumin permeability in podocytes. These negative effects were completely reversed in CatC-depleted podocytes. Moreover, PMA, but not high glucose, increased glycolytic flux in podocytes. Finally, we demonstrated that CatC expression and activity are increased in the urine of diabetic Zucker rats. We propose a novel mechanism of podocyte injury in diabetes, providing deeper insight into the role of CatC in podocyte biology.
Sigma-Aldrich
Anti-SLC2A4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Cofilin (pSer3) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Human ALB / Serum albumin ELISA Kit
Sigma-Aldrich
Anti-Nephrin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Cofilin antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture