Merck
  • Direct lineage tracing reveals Activin-a potential for improved pancreatic homing of bone marrow mesenchymal stem cells and efficient ß-cell regeneration in vivo.
Direct lineage tracing reveals Activin-a potential for improved pancreatic homing of bone marrow mesenchymal stem cells and efficient ß-cell regeneration in vivo.
Stem cell research & therapy (2020-08-01)
Nidheesh Dadheech, Abhay Srivastava, Mitul Vakani, Paresh Shrimali, Ramesh Bhonde, Sarita Gupta
ABSTRACT
Despite the potential, bone marrow-derived mesenchymal stem cells (BMSCs) show limitations for beta (ß)-cell replacement therapy due to inefficient methods to deliver BMSCs into pancreatic lineage. In this study, we report TGF-ß family member protein, Activin-a potential to stimulate efficient pancreatic migration, enhanced homing and accelerated ß-cell differentiation. Lineage tracing of permanent green fluorescent protein (GFP)- tagged donor murine BMSCs transplanted either alone or in combination with Activin-a in diabetic mice displayed potential ß-cell regeneration and reversed diabetes. Pancreatic histology of Activin-a treated recipient mice reflected high GFP+BMSC infiltration into damaged pancreas with normalized fasting blood glucose and elevated serum insulin. Whole pancreas FACS profiling of GFP+ cells displayed significant homing of GFP+BMSC with Activin-a treatment (6%) compared to BMSCs alone transplanted controls (0.5%). Within islets, approximately 5% GFP+ cells attain ß-cell signature (GFP+ Ins+) with Activin-a treatment versus controls. Further, double immunostaining for mesenchymal stem cell markers CD44+/GFP+ in infiltrated GFP+BMSC deciphers substantial endocrine reprogramming and ß-cell differentiation (6.4% Ins+/GFP+) within 15 days. Our investigation thus presents a novel pharmacological approach for stimulating direct migration and homing of therapeutic BMSCs that re-validates BMSC potential for autologous stem cell transplantation therapy in diabetes.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vimentin−Cy3 antibody produced in mouse, clone V9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Somatostatin antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Nestin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture