Merck
  • Arid1b haploinsufficiency in parvalbumin- or somatostatin-expressing interneurons leads to distinct ASD-like and ID-like behavior.
Arid1b haploinsufficiency in parvalbumin- or somatostatin-expressing interneurons leads to distinct ASD-like and ID-like behavior.
Scientific reports (2020-05-14)
Amanda L Smith, Eui-Man Jung, Byeong Tak Jeon, Woo-Yang Kim
ABSTRACT
Inhibitory interneurons are essential for proper brain development and function. Dysfunction of interneurons is implicated in several neurodevelopmental disorders, including autism spectrum disorder (ASD) and intellectual disability (ID). We have previously shown that Arid1b haploinsufficiency interferes with interneuron development and leads to social, cognitive, and emotional impairments consistent with ASD and ID. It is unclear, however, whether interneurons play a major role for the behavioral deficits in Arid1b haploinsufficiency. Furthermore, it is critical to determine which interneuron subtypes contribute to distinct behavioral phenotypes. In the present study, we generated Arid1b haploinsufficient mice in which a copy of the Arid1b gene is deleted in either parvalbumin (PV) or somatostatin (SST) interneurons, and examined their ASD- and ID-like behaviors. We found that Arid1b haploinsufficiency in PV or SST interneurons resulted in distinct features that do not overlap with one another. Arid1b haploinsufficiency in PV neurons contributed to social and emotional impairments, while the gene deletion in the SST population caused stereotypies as well as learning and memory dysfunction. These findings demonstrate a critical role of interneurons in Arid1b haploinsufficient pathology and suggest that PV and SST interneurons may have distinct roles in modulating neurological phenotypes in Arid1b haploinsufficiency-induced ASD and ID.
Sigma-Aldrich
Anti-GABA Transporter-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®