Merck
  • Direct chiral resolution of tartaric acid in food products by ligand exchange capillary electrophoresis using copper(II)-D-quinic acid as a chiral selector.
Direct chiral resolution of tartaric acid in food products by ligand exchange capillary electrophoresis using copper(II)-D-quinic acid as a chiral selector.
Journal of chromatography. A (2001-11-07)
S Kodama, A Yamamoto, A Matsunaga, K Hayakawa
ABSTRACT
Chiral resolution of native DL-tartaric acid was performed by ligand-exchange capillary electrophoresis using copper(II)-D-quinic acid as a chiral selector. Factors affecting chiral resolution, migration time, and peak area of tartaric acid were studied. The running conditions for optimum separation of tartaric acid were found to be 1 mM copper(II) sulfate-10 mM D-quinic acid (pH 5.0) with an effective voltage of -15 kV at 30 degrees C, using direct detection at 250 nm, and resolution of racemic tartaric acid was approximately 1.3. With this system, chiral resolution of DL-tartaric acid in food products was conducted successfully.
Sigma-Aldrich
D-(−)-Quinic acid, 98%