Merck
  • Assembly of Mitochondrial Complex I Requires the Low-Complexity Protein AMC1 in Chlamydomonas reinhardtii.
Assembly of Mitochondrial Complex I Requires the Low-Complexity Protein AMC1 in Chlamydomonas reinhardtii.
Genetics (2020-02-23)
Nitya Subrahmanian, Andrew David Castonguay, Claire Remacle, Patrice Paul Hamel
ABSTRACT
Complex I is the first enzyme involved in the mitochondrial electron transport chain. With >40 subunits of dual genetic origin, the biogenesis of complex I is highly intricate and poorly understood. We used Chlamydomonas reinhardtii as a model system to reveal factors involved in complex I biogenesis. Two insertional mutants, displaying a complex I assembly defect characterized by the accumulation of a 700 kDa subcomplex, were analyzed. Genetic analyses showed these mutations were allelic and mapped to the gene AMC1 (Cre16.g688900) encoding a low-complexity protein of unknown function. The complex I assembly and activity in the mutant was restored by complementation with the wild-type gene, confirming AMC1 is required for complex I biogenesis. The N terminus of AMC1 targets a reporter protein to yeast mitochondria, implying that AMC1 resides and functions in the Chlamydomonas mitochondria. Accordingly, in both mutants, loss of AMC1 function results in decreased abundance of the mitochondrial nd4 transcript, which encodes the ND4 membrane subunit of complex I. Loss of ND4 in a mitochondrial nd4 mutant is characterized by a membrane arm assembly defect, similar to that exhibited by loss of AMC1. These results suggest AMC1 is required for the production of mitochondrially-encoded complex I subunits, specifically ND4. We discuss the possible modes of action of AMC1 in mitochondrial gene expression and complex I biogenesis.
Product Description
Supelco
5-Pentadecylresorcinol, analytical standard
Sigma-Aldrich
Sodium taurodeoxycholate hydrate, ≥95% (HPLC)
Sigma-Aldrich
Myxothiazol, from Myxococcus fulvus Mx f85, ≥98% (HPLC)