Merck
  • Synergistic factors control kinase-phosphatase organization in B-cells engaged with supported bilayers.
Synergistic factors control kinase-phosphatase organization in B-cells engaged with supported bilayers.
Molecular biology of the cell (2019-12-27)
Marcos Francisco Núñez, Kathleen Wisser, Sarah L Veatch
ABSTRACT
B-cells become activated by ligands with varying valency and mode of presentation to the B-cell receptor (BCR). We previously demonstrated that clustering the immunoglobulin M (IgM) isotype of BCR with an artificial soluble cross-linker stabilized an ordered phase-like domain that enriched kinases and depleted phosphatases to promote receptor tyrosine phosphorylation. BCR is also activated by ligands presented at surfaces, and here we activate B-cells via supported bilayers of phosphatidylcholine lipids, a natural ligand for the IgM BCR expressed in the CH27 cells used. Using superresolution fluorescence localization microscopy, along with a quantitative cross-correlation analysis, we find that BRCs engaged with bilayers sort minimal peptide markers of liquid-ordered and liquid-disordered phases, indicating that ordered-domain stabilization is a general feature of BCR clustering. The phosphatase CD45 is more strongly excluded from bilayer-engaged BRCs than a transmembrane peptide, indicating that mechanisms other than domain partitioning contribute to its organization. Experimental observations are assembled into a minimal model of receptor activation that incorporates both ordered domains and direct phosphatase exclusion mechanisms to produce a more sensitive response.
Sigma-Aldrich
Atto 655 NHS ester, BioReagent, suitable for fluorescence
ALNOCHROMIX, box of 10 packs (10×3.1 oz PACKS)
Sigma-Aldrich
CH27 Mouse B Cell Lymphoma Cell Line, CH27 mouse B cell lymphoma celll line is a useful model for the study of B-cell biology and tumor immunology.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Transferrin-biotin labeled human, lyophilized powder containing sodium citrate