Merck
  • Prostaglandins and other eicosanoids in insects: biological significance.
Prostaglandins and other eicosanoids in insects: biological significance.
Annual review of entomology (2005-12-08)
David Stanley
ABSTRACT
Prostaglandins and other eicosanoids are oxygenated metabolites of certain polyunsaturated fatty acids. These compounds are well known for their important actions in mammalian physiology and disease. Recent work has revealed the presence and biological actions of eicosanoids in insects and many other invertebrate animals. In insects, eicosanoids mediate cellular immunity to microbial and metazoan challenge. Notably, some infectious organisms secrete factors responsible for impairing host insect immune reactions by inhibiting biosynthesis of eicosanoids. Eicosanoids also act in insect reproductive biology, in ion transport physiology, and in fever response to infection as well as in protein exocytosis in tick salivary glands. Aside from ongoing actions in homeostasis, certain eicosanoid actions occur at crucial points in insect life histories, such as during infectious challenge and important events in reproduction.
Product Description
Avanti
PGF1β, Avanti Research - A Croda Brand 900110P, powder
Avanti
PGE1, Avanti Research - A Croda Brand 900100P, powder